Please login to your online account to display your discounted pricing

anti-Musashi-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001759SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001759SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4440
Test Specificity Musashi 1 - Neuronal Stem Cell Marker. Reacts with human, rat, and mouse Musashi 1 protein.
Concentration 1mg/mL
Formulation PBS
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mg
Conjugate Unlabeled
Immunogen Reacts with residues 5-21 [APQPGLASPDSPHDPCK] of the human
Host Species Rabbit
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Musashi-1
Format Affinity Purified