missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Musashi-1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200 to 250 of human Musashi-1 was used as immunogen.
Antigen Musashi-1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224681 Novus Biologicals Canada
NBP224681
0.1mg Each for $257.41
NBP224681SS Novus Biologicals Canada
NBP224681SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Musashi-1
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, opossum, horse and cow, 93% homologous in chicken and Xenopus and 81% homologous in zebrafish.
4440
0.5mg/mL
A portion of amino acids 200 to 250 of human Musashi-1 was used as immunogen.
Polyclonal
Primary
Western Blot
O43347
100μg in 200μL PBS containing 0.05% BSA