Please login to your online account to display your discounted pricing

anti-Musashi-1, Polyclonal, Novus Biologicals™

$109.00 - $362.00

Specifications

Host Species Rabbit
Immunogen Reacts with residues 5-21 [APQPGLASPDSPHDPCK] of the human
Antigen Musashi-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001759

 
novus biologicals canada
NB1001759
Unlabeled 0.1mg Each for $362.00

NB1001759SS

 
novus biologicals canada
NB1001759SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Reacts with residues 5-21 [APQPGLASPDSPHDPCK] of the human
Antigen Musashi-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Musashi 1 - Neuronal Stem Cell Marker. Reacts with human, rat, and mouse Musashi 1 protein.
Gene ID (Entrez) 4440
Formulation PBS
Concentration 1mg/mL