Please login to your online account to display your discounted pricing

anti-Musashi-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224681

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224681

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4440
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, opossum, horse and cow, 93% homologous in chicken and Xenopus and 81% homologous in zebrafish.
Concentration 0.5mg/mL
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Quantity 0.1mg
Gene Accession No. O43347
Conjugate Unlabeled
Applications Western Blotting
Regulatory Status RUO
Antigen Musashi-1
Immunogen A portion of amino acids 200 to 250 of human Musashi-1 was used as immunogen.
Host Species Rabbit