Please login to your online account to display your discounted pricing

GnRH Polyclonal Antibody, Invitrogen™


GnRH Antibody, PA1-120, from Invitrogen™. Species Reactivity: Mouse; Applications: Immunofluorescence, Immunohistochemistry.

Manufacturer: pierce biotechnology inc  PA1120

Catalog No. PA1-120

  • / Each
 

Description & Specifications

Specifications

Gene Symbols GNRH1
Formulation 100μL purified antibody in PBS and 0.05% sodium azide
Gene ID (Entrez) 14714
Applications Immunofluorescence
Applications Immunohistochemistry
Storage Requirements Avoid Freeze/Thaw Cycles
Isotype IgG
Gene Alias Gonadotropin Releasing Hormone, LHRH, Luteinizing Hormone Releasing Hormone
Gene Accession No. P13562
Species Reactivity Mouse
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH.
Quantity 100μL
Antigen GnRH
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Mouse
Format Purified

GnRH Antibody, PA1-120, from Invitrogen. Species Reactivity: Mouse; Applications: Immunofluorescence, Immunohistochemistry.

Gonadotropin releasing hormone (GnRH), also known as luteinizing hormone releasing hormone (LHRH), is a key molecule in the regulation of reproduction in vertebrates. GnRH, a decapeptide, is produced by neurons in the medial basal hypothalamus (MBH) and secreted in a pulsatile manner into the cardiovascular system. The frequency and amplitude of GnRH pulses determine secretion of follicle stimulating hormone (FSH) and luteinizing hormone (LH) from the pituitary. Higher frequencies (greater than one pulse per hour) stimulate LH secretion while lower frequencies stimulate FSH secretion. The generation of GnRH pulses is effected by numerous stimuli, such as neural, hormonal and environmental. Therefore, behavioral and physiological conditions such as sleep, exercise, and stress can affect the GnRH pulses and cause a disruption of the normal cycle.   Recent studies show that GnRH also has a role in mediating cancer. GnRH has been shown to inhibit the growth of human uterine leiomyloma cells by suppressing proliferation and inducing apoptosis. GnRH analogs have been used to treat a wide variety of reproductive cancers, although the side effects of using such compounds are often quite severe.
Host Species: Rabbit
Cross Reactivity: Mouse
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH.
Formulation: 100μL purified antibody in PBS and 0.05% sodium azide
Synonyms: Gonadotropin Releasing Hormone, LHRH, Luteinizing Hormone Releasing Hormone
Uniprot Accession: P13562
Entrez GeneID: 14714
Gene Symbol: GNRH1
PA1-120 detects gonadotropin releasing hormone (GnRH) in mouse brain. PA1-120 has been successfully used in immunofluorescence and immunohistochemistry procedures. Immunofluorescent staining of GnRH in mouse embryo brain tissue using PA1-120 results in staining of neurons in the rostral forebrain. In immunohistochemical procedures, 4% paraformaldehyde is recommended as a fixative. The PA1-120 immunogen is a synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH. This peptide (Cat. # PEP-168) is available for use in neutralization and control experiments.
References:
Test: IHC (P), Dilution: 1:1000, Species: Ms, Reference: Neurokinin B is critical for normal timing of sexual maturation but dispensable for adult reproductive function in female mice. (Apr 2015) Endocrinology, Vol: 156, 4, 1386-97, True C,Nasrin Alam S,Cox K,Chan YM,Seminara SB; Pubmed ID: 25574869