missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-MTSS1, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 150-200 of human MTSS1 protein was used as the immunogen.
Antigen MTSS1
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224717 Novus Biologicals Canada
NBP224717
0.1mg Each for $307.00
NBP224717SS Novus Biologicals Canada
NBP224717SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
A portion of amino acid 150-200 of human MTSS1 protein was used as the immunogen.
Polyclonal
Primary
Western Blot
O43312
100μg in 200μL PBS containing 0.05% BSA
IgG
MTSS1
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_c, CRA_d, CRA_e, CRA_f, CRA_g) and chimpanzee, 94% homologous in monkey and 82% homologous in dog.
9788
0.5mg/mL