Please login to your online account to display your discounted pricing

anti-MTSS1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224717

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224717

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Conjugate Unlabeled
Gene ID (Entrez) 9788
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_c, CRA_d, CRA_e, CRA_f, CRA_g) and chimpanzee, 94% homologous in monkey and 82% homologous in dog.
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. O43312
Isotype IgG
Primary or Secondary Primary
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Western Blotting
Antigen MTSS1
Quantity 0.1mg
Format Purified
Immunogen A portion of amino acid 150-200 of human MTSS1 protein was used as the immunogen.
Host Species Rabbit
Regulatory Status RUO