Please login to your online account to display your discounted pricing

anti-MTH1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100109

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100109

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Concentration 1.0mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Gene Accession No. P36639
Host Species Rabbit
Immunogen A C-terminal peptide derived from human MTH1
Gene ID (Entrez) 4521
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation PBS
Regulatory Status RUO
Antigen MTH1
Quantity 0.2mL