Please login to your online account to display your discounted pricing

anti-MSP/MST1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056448SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056448T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene Accession No. Q13043
Host Species Rabbit
Applications Western Blotting
Gene ID (Entrez) 4485
Test Specificity In Jurkat, a 54 kDa band is observed. Cross reacts with Human. Predicted to cross react with Mouse and Rat based on sequence identity.
Concentration 0.5mg/mL
Antigen MSP/MST1
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 372-390 (AEDEEEEGTMKRRDETMQP) of human MST-1
Cross Reactivity Human
Format Purified
Regulatory Status RUO
Quantity 0.025mg