Please login to your online account to display your discounted pricing

anti-MSP/MST1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 372-390 (AEDEEEEGTMKRRDETMQP) of human MST-1
Antigen MSP/MST1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056448

 
novus biologicals canada
NB10056448
Unlabeled 0.1mg Each for $255.80

NB10056448T

 
novus biologicals canada
NB10056448SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 372-390 (AEDEEEEGTMKRRDETMQP) of human MST-1
Antigen MSP/MST1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In Jurkat, a 54 kDa band is observed. Cross reacts with Human. Predicted to cross react with Mouse and Rat based on sequence identity.
Gene Accession No. Q13043
Gene ID (Entrez) 4485
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL