Learn More
mitochondrial ribosomal protein S30, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MRPS30 protein.
Supplier: Abnova Corporation H00010884B01
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that is similar to the chicken pro-apoptotic protein p52. Transcript variants using alternative promoters or polyA sites have been mentioned in the literature but the complete description of these sequences is not available. [provided by RefSeq
Sequence: MAAARCWRPLLRGPRLSLHTAANAAATATETTCQDVAATPVARYPPIVASMTADSKAARLRRIERWQATVHAAESVDEKLRILTKMQFMKYMVYPQTFALNADRWYQYFTKTVFLSGLPPPPAEPEPEPEPEPEPALDLAALRAVACDCLLQEHFYLRRRRRVHRYEESEVISLPFLDQLVSTLVGLLSPHNPALAAAALDYRCPVHFYWVRGEEIIPRGHRRGRIDDLRYQIDDKPNNQIRISKQLAEFVPLDYSVPIEIPTIKCKPDKLPLFKRQYENHIFVGSKTADPCCYGHTQFHLLPDKLRRERLLRQNCADQIEVVFRANAIASLFAWTGAQAMYQGFWSEADVTRPFVSQAVITDGKYFSFFCYQLNTLALTTQADQNNPRKNICWGTQSKPLYETIEDNDVKGFNDDVLLQIVHFLLNRPKEEKSQLLENSpecifications
mitochondrial ribosomal protein S30 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MRPS30 protein. | |
MRPS30 | |
DKFZp566B2024/MRP-S30/PAP/PDCD9 | |
Mouse | |
50 μL | |
Apoptosis | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_016640 | |
MRPS30 | |
MRPS30 (NP_057724, 1 a.a. ∼ 439 a.a) full-length human protein. | |
RUO | |
Yes | |
10884 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |