Please login to your online account to display your discounted pricing

anti-XRCC3, Clone: 10F1/6, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100180

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100180

  • / Each
 

Description & Specifications

Specifications

Antigen XRCC3
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 10F1/6
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS, pH 7.4
Gene Accession No. O43542
Host Species Murine
Immunogen His-tagged human XRCC3
Isotype IgG1, κ
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7517
Test Specificity This is specific for XRCC3. NB100-180 does not cross-react with Rad51B, Rad51C, Rad51D, Rad51, or XRCC2 in Western analysis. Human. Does not work in mouse. Predicted to react with monkey based on 100% sequence homology. Immunogen has 93% homology to sheep, rat, and bovine.
Target Species Human