Please login to your online account to display your discounted pricing

anti-XRCC3, Clone: 10F1/6, Novus Biologicals™

$144.63 - $362.00

Specifications

Clone 10F1/6
Host Species Mouse
Isotype IgG1, κ
Immunogen His-tagged human XRCC3
Antigen XRCC3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100180SS

 
novus biologicals canada
NB100180SS
Unlabeled 0.025mL Each for $144.63

NB100180

 
novus biologicals canada
NB100180
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Clone 10F1/6
Host Species Mouse
Isotype IgG1, κ
Immunogen His-tagged human XRCC3
Antigen XRCC3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity This is specific for XRCC3. NB100-180 does not cross-react with Rad51B, Rad51C, Rad51D, Rad51, or XRCC2 in Western analysis. Human. Does not work in mouse. Predicted to react with monkey based on 100% sequence homology. Immunogen has 93% homology to sheep, rat, and bovine.
Gene Accession No. O43542
Gene ID (Entrez) 7517
Formulation PBS, pH 7.4
Concentration 1mg/mL