Please login to your online account to display your discounted pricing

anti-XRCC3, Clone: 10F1/6, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100180SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100180SS

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen XRCC3
Formulation PBS, pH 7.4
Quantity 0.025mL
Isotype IgG1, κ
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Gene ID (Entrez) 7517
Concentration 1mg/mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This is specific for XRCC3. NB100-180 does not cross-react with Rad51B, Rad51C, Rad51D, Rad51, or XRCC2 in Western analysis. Human. Does not work in mouse. Predicted to react with monkey based on 100% sequence homology. Immunogen has 93% homology to sheep, rat, and bovine.
Immunogen His-tagged human XRCC3
Gene Accession No. O43542
Conjugate Unlabeled
Clone 10F1/6
Cross Reactivity Human