Please login to your online account to display your discounted pricing

anti-Wnt-5a, Clone: 4M5E4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227295SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227295SS

  • / Each
 

Description & Specifications

Specifications

Antigen Wnt-5a
Applications Immunohistochemistry (Paraffin)
Clone 4M5E4
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Gene Accession No. P41221
Host Species Murine
Immunogen A portion of the human Wnt5A protein was used as the immunogen for this antibody.
Isotype κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7474
Target Species Human