Please login to your online account to display your discounted pricing

anti-Wnt-5a, Clone: 4M5E4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227295

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227295

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7474
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Gene Accession No. P41221
Applications Immunohistochemistry (Paraffin)
Primary or Secondary Primary
Isotype κ
Format Purified
Quantity 0.1mg
Clone 4M5E4
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Host Species Mouse
Immunogen A portion of the human Wnt5A protein was used as the immunogen for this antibody.
Regulatory Status RUO
Antigen Wnt-5a