Please login to your online account to display your discounted pricing

anti-Wnt-5a, Clone: 4M5E4, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 4M5E4
Host Species Murine
Isotype κ
Immunogen A portion of the human Wnt5A protein was used as the immunogen for this antibody.
Antigen Wnt-5a
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227295

 
novus biologicals canada
NBP227295
Unlabeled 0.1mg Each for $309.05

NBP227295SS

 
novus biologicals canada
NBP227295SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 4M5E4
Host Species Murine
Isotype κ
Immunogen A portion of the human Wnt5A protein was used as the immunogen for this antibody.
Antigen Wnt-5a
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Gene Accession No. P41221
Gene ID (Entrez) 7474
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL