Please login to your online account to display your discounted pricing

anti-Von Willebrand Factor, Clone: 3E2D10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233002

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233002

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Host Species Mouse
Immunogen Recombinant human vWF fragment spanning aa 845-949
Format Purified
Quantity 0.5mL
Clone 3E2D10
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Gene ID (Entrez) 7450
Gene Accession No. P04275
Concentration 0.2mg/mL
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Isotype κ
Cross Reactivity Human
Antigen Von Willebrand Factor