Please login to your online account to display your discounted pricing

anti-Von Willebrand Factor, Clone: 3E2D10, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 3E2D10
Host Species Mouse
Isotype κ
Immunogen Recombinant human vWF fragment spanning aa 845-949
Antigen Von Willebrand Factor
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2330021M

 
novus biologicals canada
NBP2330021ML
Unlabeled 1mL Each for $429.00

NBP233002

 
novus biologicals canada
NBP233002
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone 3E2D10
Host Species Mouse
Isotype κ
Immunogen Recombinant human vWF fragment spanning aa 845-949
Antigen Von Willebrand Factor
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P04275
Gene ID (Entrez) 7450
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL