Please login to your online account to display your discounted pricing

anti-Von Willebrand Factor, Clone: 3E2D10, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2330021ML

 View all options for this product


For Research Use Only

Catalog No. NBP2330021M


Description & Specifications


Antigen Von Willebrand Factor
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone 3E2D10
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04275
Host Species Mouse
Immunogen Recombinant human vWF fragment spanning aa 845-949
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7450