Please login to your online account to display your discounted pricing

anti-Vimentin, Clone: 2D1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP192687SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP192687SS

  • / Each
 

Description & Specifications

Specifications

Antigen Vimentin
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Clone 2D1
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS
Gene Accession No. P08670
Host Species Murine
Immunogen Recombinant human Vimentin expressed in and purified from &italic_begin;E. coli&italic_end;. [UniProt No. P08670]
Isotype κ
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7431
Test Specificity Human, Rat and Mouse and likely all other Mammals.
Target Species Human
Target Species Mammalia
Target Species Murine
Target Species Rat