Please login to your online account to display your discounted pricing

anti-Vimentin, Clone: 2D1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP192687

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP192687

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mL
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Gene Accession No. P08670
Isotype κ
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Primary or Secondary Primary
Concentration 1mg/mL
Gene ID (Entrez) 7431
Format Purified
Test Specificity Human, Rat and Mouse and likely all other Mammals.
Formulation PBS
Immunogen Recombinant human Vimentin expressed in and purified from &italic_begin;E. coli&italic_end;. [UniProt No. P08670]
Host Species Mouse
Antigen Vimentin
Clone 2D1