Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, Clone: KDR721, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone KDR721
Host Species Mouse
Isotype κ
Immunogen Recombinant human VEGF-R2 protein
Antigen VEGF R2/KDR/Flk-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233001

 
novus biologicals canada
NBP233001
Unlabeled 0.5mL Each for $321.00

NBP2330011M

 
novus biologicals canada
NBP2330011ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone KDR721
Host Species Mouse
Isotype κ
Immunogen Recombinant human VEGF-R2 protein
Antigen VEGF R2/KDR/Flk-1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene Accession No. P35968
Gene ID (Entrez) 3791
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL