Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, Clone: KDR721, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2330011ML

 View all options for this product


For Research Use Only

Catalog No. NBP2330011M


Description & Specifications


Antigen VEGF R2/KDR/Flk-1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone KDR721
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P35968
Host Species Mouse
Immunogen Recombinant human VEGF-R2 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3791