Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, Clone: KDR657, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233000

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233000

  • / Each
 

Description & Specifications

Specifications

Antigen VEGF R2/KDR/Flk-1
Applications ELISA
Clone KDR657
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P35968
Host Species Mouse
Immunogen Recombinant human VEGF-R2 protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3791