Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, Clone: KDR657, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone KDR657
Host Species Mouse
Isotype κ
Immunogen Recombinant human VEGF-R2 protein
Antigen VEGF R2/KDR/Flk-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233000

 
novus biologicals canada
NBP233000
Unlabeled 0.5mL Each for $321.00

NBP2330001M

 
novus biologicals canada
NBP2330001ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone KDR657
Host Species Mouse
Isotype κ
Immunogen Recombinant human VEGF-R2 protein
Antigen VEGF R2/KDR/Flk-1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Gene Accession No. P35968
Gene ID (Entrez) 3791
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL