Please login to your online account to display your discounted pricing

anti-VEGF R2/KDR/Flk-1, Clone: KDR657, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2330001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2330001M

  • / Each
 

Description & Specifications

Specifications

Format Purified
Cross Reactivity Human
Antigen VEGF R2/KDR/Flk-1
Isotype κ
Formulation PBS with 0.05% BSA.
Immunogen Recombinant human VEGF-R2 protein
Regulatory Status RUO
Conjugate Unlabeled
Gene Accession No. P35968
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 3791
Concentration 0.2mg/mL
Host Species Mouse
Applications ELISA
Quantity 1mL
Clone KDR657