Please login to your online account to display your discounted pricing

anti-VEGF R1/Flt-1, Clone: FLT1/659, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2329091ML

 View all options for this product


For Research Use Only

Catalog No. NBP2329091M


Description & Specifications


Antigen VEGF R1/Flt-1
Applications ELISA
Clone FLT1/659
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P17948
Host Species Mouse
Immunogen Recombinant human VEGF-R1 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2321