Please login to your online account to display your discounted pricing

anti-VEGF R1/Flt-1, Clone: FLT1/659, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232909

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232909

  • / Each
 

Description & Specifications

Specifications

Antigen VEGF R1/Flt-1
Host Species Mouse
Applications ELISA
Immunogen Recombinant human VEGF-R1 protein
Isotype κ
Quantity 0.5mL
Clone FLT1/659
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Conjugate Unlabeled
Gene Accession No. P17948
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2321
Concentration 0.2mg/mL
Cross Reactivity Human
Format Purified