Please login to your online account to display your discounted pricing

anti-VEGF R1/Flt-1, Clone: FLT1/659, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone FLT1/659
Host Species Mouse
Isotype κ
Immunogen Recombinant human VEGF-R1 protein
Antigen VEGF R1/Flt-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232909

 
novus biologicals canada
NBP232909
Unlabeled 0.5mL Each for $321.00

NBP2329091M

 
novus biologicals canada
NBP2329091ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone FLT1/659
Host Species Mouse
Isotype κ
Immunogen Recombinant human VEGF-R1 protein
Antigen VEGF R1/Flt-1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Gene Accession No. P17948
Gene ID (Entrez) 2321
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL