Please login to your online account to display your discounted pricing

anti-VEGF R1/Flt-1, Clone: FLT1/658, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232908

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232908

  • / Each
 

Description & Specifications

Specifications

Antigen VEGF R1/Flt-1
Applications ELISA
Clone FLT1/658
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P17948
Host Species Mouse
Immunogen Recombinant human VEGF-R1 protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2321