Please login to your online account to display your discounted pricing

anti-VEGF R1/Flt-1, Clone: FLT1/658, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone FLT1/658
Host Species Mouse
Isotype κ
Immunogen Recombinant human VEGF-R1 protein
Antigen VEGF R1/Flt-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329081M

 
novus biologicals canada
NBP2329081ML
Unlabeled 1mL Each for $429.00

NBP232908

 
novus biologicals canada
NBP232908
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone FLT1/658
Host Species Mouse
Isotype κ
Immunogen Recombinant human VEGF-R1 protein
Antigen VEGF R1/Flt-1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Gene Accession No. P17948
Gene ID (Entrez) 2321
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL