Please login to your online account to display your discounted pricing

anti-VEGF, Clone: MM0009-2B86, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11060975SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11060975T

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen VEGF
Clone MM0009-2B86
Format Purified
Quantity 0.02mg
Cross Reactivity Human
Host Species Mouse
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human VEGF-165 recombinant protein).
Monoclonal or Polyclonal Monoclonal
Reconstitution Reconstitute the antibody with 500μL of sterile PBS to a final concentration of 200μg/mL.
Gene ID (Entrez) 7422
Test Specificity This antibody detects both human VEGF 165 and 121.
Formulation Lyophilized.
Conjugate Unlabeled
Primary or Secondary Primary
Regulatory Status RUO
Gene Accession No. P15692