Please login to your online account to display your discounted pricing

anti-VEGF, Clone: MM0009-2B86, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11060975

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11060975

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Host Species Mouse
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human VEGF-165 recombinant protein).
Quantity 0.1mg
Regulatory Status RUO
Clone MM0009-2B86
Reconstitution Reconstitute the antibody with 500μL of sterile PBS to a final concentration of 200μg/mL.
Conjugate Unlabeled
Test Specificity This antibody detects both human VEGF 165 and 121.
Formulation Lyophilized.
Gene Accession No. P15692
Gene ID (Entrez) 7422
Antigen VEGF
Format Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Isotype IgG2