Please login to your online account to display your discounted pricing

anti-VEGF, Clone: MM0009-2B86, Novus Biologicals™

$110.17 - $387.37

Specifications

Clone MM0009-2B86
Host Species Murine
Isotype IgG2
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human VEGF-165 recombinant protein).
Antigen VEGF
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11060975

 
novus biologicals canada
NB11060975
Unlabeled 0.1mg Each for $387.37

NB11060975T

 
novus biologicals canada
NB11060975SS
Unlabeled 0.02mg Each for $110.17
Description & Specifications

Specifications

Clone MM0009-2B86
Host Species Murine
Isotype IgG2
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human VEGF-165 recombinant protein).
Antigen VEGF
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This antibody detects both human VEGF 165 and 121.
Gene Accession No. P15692
Gene ID (Entrez) 7422
Formulation Lyophilized.
Reconstitution Reconstitute the antibody with 500μL of sterile PBS to a final concentration of 200μg/mL.