Please login to your online account to display your discounted pricing

anti-VABP, Clone: 6C239.4, Novus Biologicals™

$139.00 - $259.02

Specifications

Clone 6C239.4
Host Species Mouse
Isotype κ
Immunogen Full length recombinant protein was used as the immunogen.
Antigen VABP
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224876SS

 
novus biologicals canada
NBP224876SS
Unlabeled 0.025mg Each for $139.00

NBP224876

 
novus biologicals canada
NBP224876
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Clone 6C239.4
Host Species Mouse
Isotype κ
Immunogen Full length recombinant protein was used as the immunogen.
Antigen VABP
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity This antibody is reactive to Human.
Gene Accession No. Q9NZI2
Gene ID (Entrez) 30820
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL