Please login to your online account to display your discounted pricing

anti-TYRP1, Clone: TA99, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232906

 View all options for this product


For Research Use Only

Catalog No. NBP232906


Description & Specifications


Antigen TYRP1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone TA99
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P17643
Host Species Mouse
Immunogen SK-MEL-23 cells
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7306