Please login to your online account to display your discounted pricing

anti-TYRP1, Clone: TA99, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone TA99
Host Species Mouse
Isotype κ
Immunogen SK-MEL-23 cells
Antigen TYRP1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232906

 
novus biologicals canada
NBP232906
Unlabeled 0.5mL Each for $321.00

NBP2329061M

 
novus biologicals canada
NBP2329061ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone TA99
Host Species Mouse
Isotype κ
Immunogen SK-MEL-23 cells
Antigen TYRP1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P17643
Gene ID (Entrez) 7306
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL