Please login to your online account to display your discounted pricing

anti-TTF1 / NKX2.1, Clone: 8G7G3/1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP229434

 View all options for this product


For Research Use Only

Catalog No. NBP229434


Description & Specifications


Antigen TTF1 / NKX2.1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone 8G7G3/1
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Formulation Culture Medium
Gene Accession No. P43699
Host Species Mouse
Immunogen A full length recombinant rat TTF-1 protein was used as the immunogen for the 8G7G3/1 antibody.
Isotype IgG1
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7080
Test Specificity Recognizes a protein of 40kDa, identified as Thyroid transcription factor-1 (TTF-1). TTF-1 is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. Anti-TTF-1 is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the breast, mediastinal germ cell tumors, and malignant mesothelioma. It can also be used to differentiate small cell lung carcinoma from lymphoid infiltrates. Loss of TTF-1 expression in non-small cell lung carcinoma has been associated with aggressive behavior of such neoplasms. TTF-1 reactivity is also seen in thyroid malignancies.