Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Clone: 59N5G4, Novus Biologicals™

$139.00 - $259.02

Specifications

Clone 59N5G4
Host Species Mouse
Immunogen Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody.
Antigen TSLP R/CRLF2
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227331

 
novus biologicals canada
NBP227331
Unlabeled 0.1mg Each for $259.02

NBP227331SS

 
novus biologicals canada
NBP227331SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 59N5G4
Host Species Mouse
Immunogen Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody.
Antigen TSLP R/CRLF2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity TSLPR is a glycoprotein with two major isoforms of 359 amino acids and 255 amino acids. In western blotting, bands may be visualized between 27 kD and ∽50 kD.
Gene Accession No. Q9HC73
Gene ID (Entrez) 64109
Formulation 0.1mg TSLPR antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL