Please login to your online account to display your discounted pricing

anti-TSLP, Clone: 55N2D4, Novus Biologicals™

$139.00 - $257.41

Specifications

Clone 55N2D4
Host Species Murine
Immunogen amino acids (27-158) MDYDFTNCDFEkIkAAYLSTISkDLITYMSGTkSTEFNNTVSCSNRPHCLTEIQSLTFNP~TAGCASLAkEMFAMkTkAALAIWCPGYSETQINATQAMkkRRkRkVTTNkCLEQVSQLQG~LWRRFNRPLLkQQL
Antigen TSLP
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227325

 
novus biologicals canada
NBP227325
Unlabeled 0.1mg Each for $257.41

NBP227325SS

 
novus biologicals canada
NBP227325SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 55N2D4
Host Species Murine
Immunogen amino acids (27-158) MDYDFTNCDFEkIkAAYLSTISkDLITYMSGTkSTEFNNTVSCSNRPHCLTEIQSLTFNP~TAGCASLAkEMFAMkTkAALAIWCPGYSETQINATQAMkkRRkRkVTTNkCLEQVSQLQG~LWRRFNRPLLkQQL
Antigen TSLP
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q969D9
Gene ID (Entrez) 85480
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL