Please login to your online account to display your discounted pricing

anti-TSLP, Biotin, Clone: 210H1.04, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0804B

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0804B

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 85480
Cross Reactivity Human
Applications ELISA
Applications Western Blotting
Antigen TSLP
Isotype IgG2a
Format Ion Exchange Chromatography
Quantity 0.1mg
Host Species Mouse
Conjugate Biotin
Formulation Antibody in PBS with 50% glycerol
Clone 210H1.04
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Immunogen human TSLP-His transfected 293T
Regulatory Status RUO