Please login to your online account to display your discounted pricing

anti-TSLP, Biotin, Clone: 210H1.04, Novus Biologicals™

$406.00 - $407.76

Specifications

Clone 210H1.04
Host Species Murine
Isotype IgG2a
Immunogen human TSLP-His transfected 293T
Antigen TSLP
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX0804B005

 
novus biologicals canada
DDX0804B005MG
Biotin 0.05mg Each for $406.00

DDX0804B

 
novus biologicals canada
DDX0804B
Biotin 0.1mg Each for $407.76
Description & Specifications

Specifications

Clone 210H1.04
Host Species Murine
Isotype IgG2a
Immunogen human TSLP-His transfected 293T
Antigen TSLP
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blot
Gene ID (Entrez) 85480
Formulation Antibody in PBS with 50% glycerol