Please login to your online account to display your discounted pricing

anti-TSLP, Biotin, Clone: 210H1.04, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0804B005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0804B005

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 85480
Regulatory Status RUO
Cross Reactivity Human
Antigen TSLP
Format Ion Exchange Chromatography
Formulation Antibody in PBS with 50% glycerol
Host Species Mouse
Applications ELISA
Applications Western Blotting
Immunogen human TSLP-His transfected 293T
Isotype IgG2a
Quantity 0.05mg
Monoclonal or Polyclonal Monoclonal
Conjugate Biotin
Primary or Secondary Primary
Clone 210H1.04