Please login to your online account to display your discounted pricing

anti-Troponin I Type 3 (cardiac), Clone: 2C9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP235336SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP235336SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Gene Accession No. P19429
Formulation PBS
Gene ID (Entrez) 7137
Concentration 1.0mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Mouse
Immunogen Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Isotype κ
Antigen Troponin I Type 3 (cardiac)
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Quantity 0.025mg
Clone 2C9