Please login to your online account to display your discounted pricing

anti-Troponin I Type 3 (cardiac), Clone: 2C9, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 2C9
Host Species Mouse
Isotype κ
Immunogen Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Antigen Troponin I Type 3 (cardiac)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP235336

 
novus biologicals canada
NBP235336
Unlabeled 0.1mg Each for $309.05

NBP235336SS

 
novus biologicals canada
NBP235336SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 2C9
Host Species Mouse
Isotype κ
Immunogen Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Antigen Troponin I Type 3 (cardiac)
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P19429
Gene ID (Entrez) 7137
Formulation PBS
Concentration 1.0mg/mL