Please login to your online account to display your discounted pricing

anti-Troponin I Type 1 (slow skeletal), Clone: 12F10, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200431

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200431

  • / Each
 

Description & Specifications

Specifications

Applications Immunoassay
Applications Sandwich ELISA
Applications Western Blotting
Host Species Mouse
Isotype IgG2b
Cross Reactivity Human
Immunogen Full length native protein (purified) (Human).
Antigen Troponin I Type 1 (slow skeletal)
Formulation PBS, pH 7.4
Gene Accession No. P19237
Primary or Secondary Primary
Gene ID (Entrez) 7135
Test Specificity Troponin I from human skeletal muscle. Cross-reaction of this with cardiac troponin I is less then 0.05%. Not yet tested in other species.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Regulatory Status RUO
Quantity 0.1mg
Format Purified
Clone 12F10