Please login to your online account to display your discounted pricing

anti-Troponin I Type 1 (slow skeletal), Clone: 12F10, Novus Biologicals™

$329.00 - $329.00

Specifications

Clone 12F10
Host Species Murine
Isotype IgG2b
Immunogen Full length native protein (purified) (Human).
Antigen Troponin I Type 1 (slow skeletal)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB20043105

 
novus biologicals canada
NB20043105MG
Unlabeled 0.5mg Each for $329.00

NB200431

 
novus biologicals canada
NB200431
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Clone 12F10
Host Species Murine
Isotype IgG2b
Immunogen Full length native protein (purified) (Human).
Antigen Troponin I Type 1 (slow skeletal)
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoassay
Applications Sandwich ELISA
Applications Western Blot
Test Specificity Troponin I from human skeletal muscle. Cross-reaction of this with cardiac troponin I is less then 0.05%. Not yet tested in other species.
Gene Accession No. P19237
Gene ID (Entrez) 7135
Formulation PBS, pH 7.4