Please login to your online account to display your discounted pricing

anti-Troponin I Type 1 (slow skeletal), Clone: 12F10, Novus Biologicals™


Manufacturer: novus biologicals canada  NB20043105MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB20043105

  • / Each
 

Description & Specifications

Specifications

Antigen Troponin I Type 1 (slow skeletal)
Host Species Mouse
Applications Immunoassay
Applications Sandwich ELISA
Applications Western Blotting
Isotype IgG2b
Regulatory Status RUO
Format Purified
Immunogen Full length native protein (purified) (Human).
Quantity 0.5mg
Formulation PBS, pH 7.4
Gene Accession No. P19237
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7135
Test Specificity Troponin I from human skeletal muscle. Cross-reaction of this with cardiac troponin I is less then 0.05%. Not yet tested in other species.
Conjugate Unlabeled
Cross Reactivity Human
Clone 12F10