Please login to your online account to display your discounted pricing

anti-TRF-2, Clone: 3H6B4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP226298SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP226298SS

  • / Each
 

Description & Specifications

Specifications

Antigen TRF-2
Applications Immunohistochemistry
Applications Western Blot
Clone 3H6B4
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Precipitation
Formulation PBS, 0.5% protein stabilizer
Gene Accession No. Q15554
Host Species Murine
Immunogen Partial recombinant human TRF2 (between residues 300-500) expressed in &italic_begin;E. coli&italic_end; [Uniprot Q15554]
Isotype IgG1
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7014
Target Species Human